Learn More
Merck MultiScreen™ Multiple Punch and Punch Tips
Shop All Merck Products
Click to view available options
Product Type:
Punch kit
Punch tip
Unit Size:
Pack of 10
Pack of 50
Description
- Punch and tips are used to simultaneously remove filters from eight wells of a MultiScreen plate and distribute them into eight scintillation vials for scintillation or gamma radioscope detection
- Remove filters from wells separately, eliminating cross-contamination
- Usable with all MultiScreen plates containing Durapore (PVDF) membrane; mixed cellulose ester membranes (HA) and glass fiber membranes must be wet when punched from plate
- Multiple punch includes punch with punch carrier plate and punch distributor
Order Info
Disposable punch tips for the multiple punch are available. Three punch kits are available—all include multiple punch and carrier racks
Specifications
Specifications
| Product Type | Punch kit |
| For Use With (Application) | Radionucleotide counting by transfer of individual filters to tubes |
| Includes | Punch with punch carrier plate and punch distributor |
| Quantity | 1/Pk. |