missing translation for 'onlineSavingsMsg'
Learn More

Merck MultiScreen™HTS 8-Well Strips with Immobilon™-P PVDF Membrane

Product Code. 10169794 Shop All Merck Products
Change view
Click to view available options
Product Type:
Microplate 8-Well Strip
Microplate Strip Support Frame
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
10169794 Microplate 8-Well Strip Pack of 10
10174073 Microplate Strip Support Frame Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10169794 Supplier Merck Supplier No. M8IPS4510
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

Twelve 8-well strips in a support frame. Additional support frames also available separately.

8-well strips enable processing of less than 96 samples at one time

  • For ELISpot and protein binding assays

TRUSTED_SUSTAINABILITY

Specifications

Format Strip Plate
Array 1 x 8
Color Clear
Product Type Microplate 8-Well Strip
Material ABS Plastic
No. of Wells 8
Sterility Sterile
For Use With (Application) ELIspot and protein binding assays
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.