missing translation for 'onlineSavingsMsg'
Learn More

Merck Pellicon™ XL Devices

Small-volume tangential flow filter device

Brand:  Merck PXB005A50

Code : 72

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11943926

  • 975.00 € / Each
Estimated Shipment: 12-07-2024
to see stock.
Add to basket

Includes: Operating instructions and Certificate of Quality

Description

Description

Designed for process development and sample preparation, with an optimal processing volume range of 50 to 1000mL.

  • True linear scalability with same flow path, channel length, and channel height as other established Pellicon products
  • Can operate with peristaltic pump or Labscale™ TFF system Robust thermoplastic (polypropylene) construction
  • Controlled channel height
  • Choice of Biomax™ PES (polyethersulfone) or Ultracel™ PLC (composite regenerated cellulose) membranes

Concentration and desalting of proteins, buffer exchange, depyrogenation, clarification, and preparation of material for clinical trials.

Order Info

Packaged one per box.

Specifications

Specifications

Ultrafiltration Biomax-5
Polypropylene
100 μm
Protein purification, analytical sample preparation
80psi at 50°C
1 Pack
Pellicon XL Ultrafiltration Device
3.2 mL
7.4 x 1.2 in. (18.8 x 3cm)
18.8 cm
3 cm
Cassette
5 kDa
50 cm2
Operating instructions and certificate of quality
1 to 14
0.03 to 0.05L/min.
0.5 oz.
15 mL
7.4 in.
1.2 in.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers