missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Emerin Antibody (CL0203) [Alexa Fluor™ 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52877AF405
This item is not returnable.
View return policy
Description
Emerin Monoclonal antibody specifically detects Emerin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
Specifications
| Emerin | |
| Monoclonal | |
| Alexa Fluor 405 | |
| emerin, Emery-Dreifuss muscular dystrophy, LEM domain containing 5, STAEDMDLEMD5 | |
| This antibody was developed using a recombinant protein derived from P50402, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cell Biology, Cytoskeleton Markers, Signal Transduction | |
| 2010 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence | |
| CL0203 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction