Learn More
Emerin Antibody (CL0203) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52877PCP
Product Code. 30296289
  • 770.00 € / 0.10mL
Description
Emerin Monoclonal antibody specifically detects Emerin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Emerin
Monoclonal
PerCP
emerin, Emery-Dreifuss muscular dystrophy, LEM domain containing 5, STAEDMDLEMD5
This antibody was developed using a recombinant protein derived from P50402, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cell Biology, Cytoskeleton Markers, Signal Transduction
2010
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence
CL0203
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified