missing translation for 'onlineSavingsMsg'
Learn More

EMILIN2 Rabbit anti-Human, Polyclonal, Proteintech

Product Code. p-7210481 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16863265 20 μL 20µL
16853265 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16863265 Supplier Proteintech Supplier No. 247791AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Specifications

Antigen EMILIN2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.49 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene EMILIN2
Gene Accession No. Q9BXX0
Gene Alias EMILIN 2, EMILIN2, FOAP 10, Protein FOAP 10
Gene Symbols EMILIN2
Host Species Rabbit
Immunogen EMILIN2 Fusion Protein Ag20722
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84034
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.