missing translation for 'onlineSavingsMsg'
Learn More

Emodin, MedChemExpress

Product Code. 16472187
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
Unit Size:
100mg
200mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472187 50 mg 50mg
16436456 100 mg 100mg
16421126 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16472187 Supplier Medchem Express Supplier No. HY14393
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Emodin (Frangula emodin), an anthraquinone derivative, is an anti-SARS-CoV compound.

Emodin blocks the SARS coronavirus spike protein and angiotensin-converting enzyme 2 (ACE2) interaction. Emodin inhibits casein kinase-2 (CK2). Anti-inflammatory and anticancer effects. Emodin is a potent selective 11β-HSD1 inhibitor with the IC50 of 186 and 86 nM for human and mouse 11β-HSD1, respectively. Emodin ameliorates metabolic disorder in diet-induced obese mice.

Specifications

Chemical Name or Material Emodin
CAS 518-82-1
Color Yellow
Purity Grade Notes Research
Molecular Formula C15H10O5
Quantity 50 mg
Synonym Frangula emodin
Solubility Information Acetone : 10.87 mg/mL (40.22 mM; Need ultrasonic) ∣DMSO : 5.41 mg/mL (20.02 mM; Need ultrasonic) ∣Ethanol : < 1 mg/mL (ultrasonic) (insoluble)
SMILES O=C1C2=C(C=C(C)C=C2O)C(C3=CC(O)=CC(O)=C31)=O
Molecular Weight (g/mol) 270.24
Formula Weight 270.24
Percent Purity 96.53%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.