missing translation for 'onlineSavingsMsg'
Learn More

Emodin, TRC

Product Code. 30007262
Change view
Click to view available options
Quantity:
25 mg
50 mg
100 mg
Unit Size:
100mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30007262 50 mg 50mg
30004058 25 mg 25mg
30006242 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007262 Supplier Toronto Research Chemicals Supplier No. TRCE52300050MG
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 518-82-1
Molecular Formula C15 H10 O5
Molecular Weight (g/mol) 270.24
Synonym 9,10-Anthracenedione, 1,3,8-trihydroxy-6-methyl-, Anthraquinone, 1,3,8-trihydroxy-6-methyl- (8CI), 1,3,8-Trihydroxy-6-methyl-9,10-anthracenedione, 1,3,8-Trihydroxy-6-methyl-9,10-anthraquinone, 1,3,8-Trihydroxy-6-methylanthraquinone, 1,6,8-Trihydroxy-3-methylanthraquinone, 3-Methyl-1,6,8-trihydroxyanthraquinone, 4,5,7-Trihydroxy-2-methylanthraquinone, Archin, Emodin, Emodol, Frangula emodin, Frangulic acid, NSC 408120, NSC 622947, Rheum emodin, Schuttgelb
IUPAC Name 1,3,8-trihydroxy-6-methylanthracene-9,10-dione
SMILES Cc1cc(O)c2C(=O)c3c(O)cc(O)cc3C(=O)c2c1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C15H10O5/c1-6-2-8-12(10(17)3-6)15(20)13-9(14(8)19)4-7(16)5-11(13)18/h2-5,16-18H,1H3
Formula Weight 270.0528
Percent Purity >95%
Chemical Name or Material Emodin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.