missing translation for 'onlineSavingsMsg'
Learn More

Emtricitabine, MedChemExpress

Product Code. 16471856
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16471856 500 mg 500mg
16433357 50 mg 50mg
16417306 100 mg 100mg
16401266 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16471856 Supplier Medchem Express Supplier No. HY17427
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Emtricitabine is a nucleoside reverse transcriptase inhibitor (NRTI) with an EC50 of 0.01 μM in PBMC cell. It is an antiviral drug for the treatment of HIV infection.

Emtricitabine is a nucleoside reverse transcriptase inhibitor (NRTI) with an EC50 of 0.01 μM in PBMC cell. It is an antiviral drug for the treatment of HIV infection.

Specifications

Chemical Name or Material Emtricitabine
Physical Form Solid
CAS 143491-57-0
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C8H10FN3O3S
For Use With (Application) COVID-19-anti-virus
Synonym BW1592
Solubility Information DMSO : ≥ 100 mg/mL (404.45 mM)
SMILES O=C1N=C(N)C(F)=CN1[C@@H]2CS[C@H](CO)O2
Molecular Weight (g/mol) 247.25
Formula Weight 247.25
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.