missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EN 14039/ISO 16703 Hydrocarbon Standard in n-Heptane, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Diesel fuel No.2∣Mineral Oil (without additives)∣n-Decane∣Tetracontane |
| CAS | 68476-34-6, 8042-47-5, 124-18-5, 4181-95-7 |
| Analyte or Component Names | Diesel fuel No.2∣Mineral Oil (without additives)∣n-Decane∣Tetracontane |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Heptane |
| Industry Type | Petrochemical Testing |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | n-Decane 0.02985 μg/mL∣Tetracontane 30 μg/mL∣Diesel fuel No.2 4000 μg/mL∣Mineral Oil (without additives) 4004 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?