missing translation for 'onlineSavingsMsg'
Learn More

EN 14039/ISO 16703/ISO 9377 n-Alkanes System Performance Standard 50 μg/mL in n-Heptane, Dr. Ehrenstorfer

Product Code. 30328574
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30328574 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30328574 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000971HP
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material n-Decane∣n-Docosane∣n-Dodecane∣n-Dotriacontane∣n-Eicosane∣n-Hexacosane∣n-Hexadecane∣n-Hexatriacontane∣n-Octadecane∣n-Octatriacontane∣n-Tetradecane∣n-Tetratriacontane∣Octacosane∣Tetracontane∣Tetracosane∣Triacontane
CAS 124-18-5, 629-97-0, 112-40-3, 544-85-4, 112-95-8, 630-01-3
Analyte or Component Names n-Decane∣n-Docosane∣n-Dodecane∣n-Dotriacontane∣n-Eicosane∣n-Hexacosane∣n-Hexadecane∣n-Hexatriacontane∣n-Octadecane∣n-Octatriacontane∣n-Tetradecane∣n-Tetratriacontane∣Octacosane∣Tetracontane∣Tetracosane∣Triacontane
Shipping Condition Ice pack (-20°C)
Certifications/Compliance ISO 17034
Solvent or Matrix Heptane
Industry Type Petrochemical Testing
Quantity 1 mL
Concentration or Composition (by Analyte or Components) n-Decane 50 μg/mL∣n-Docosane 50 μg/mL∣n-Dodecane 50 μg/mL∣n-Dotriacontane 50 μg/mL∣n-Eicosane 50 μg/mL∣n-Hexacosane 50 μg/mL∣n-Hexadecane 50 μg/mL∣n-Hexatriacontane 50 μg/mL∣n-Octadecane 50 μg/mL∣n-Octatriacontane 50 μg/mL∣n-Tetradecane 50 μg/mL∣n-Tetratriacontane 50 μg/mL∣Octacosane 50 μg/mL∣Tetracontane 50 μg/mL∣Tetracosane 50 μg/mL∣Triacontane 50 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.