missing translation for 'onlineSavingsMsg'
Learn More

R&DSystems™ Endothelial Cell Growth Base Media

Product Code. 18737367
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18737367 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18737367 Supplier R&DSystems™ Supplier No. 390598
  • ONLINE EXCLUSIVE PRICE
    92.00 €
    87.15 € / 250mL
    Save 4.85 €
    5% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

Base Media for the Low-serum Expansion of Primary Endothelial Cells

TRUSTED_SUSTAINABILITY

Specifications

Quantity 250 mL
Species Human
Content And Storage Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.