missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Endothelial Cell Growth Media

Product Code. 18710328
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
250mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18710328 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18710328 Supplier R&D Systems™ Supplier No. CCM027
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Media for the low-serum expansion of primary endothelial cells.

Endothelial Cell Base Media plus Supplement

Primary endothelial cells are commonly used for the study of developmental and disease mechanisms involving the blood vessels, such as angiogenesis. Endothelial cells are phenotypically characterized as CD31+, kDaR+, and vWF+ cells. Media has been tested for its ability to support CD31+, kDaR+, and vWF+ human umbilical vein endothelial cell growth in vitro.

  • Unique and optimized formulation that promotes endothelial cell growth
  • Reduces cell senescence at higher passages
  • Includes single supplement featuring key growth factors and small molecules

Storage: Store under sterile conditions at 2° to 8°C

TRUSTED_SUSTAINABILITY

Specifications

Product Type Growth Medium
Quantity 250 mL
Content And Storage Endothelial cell growth base media, 50X Endothelial cell growth supplement. Store the unopened product at 2 - 8° C. Do not use past expiration date.

For Laboratory research use only. Not fro use in diagnostic procedures

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.