missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ SingleQuots™ Endothelial Cell Growth Medium-2MV

Product Code. 11645500
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11645500 1 Kit Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11645500 Supplier Lonza™ Supplier No. CC4147
 Restricted Item
Estimated Shipment: 21-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Developed for the enhanced growth of lung microvascular cells. Lonza™ SingleQuots™ Endothelial Cell Growth Medium-2MV features a special formulation that is optimized to grow human coronary and iliac artery endothelial cells. Guaranteed to perform when used with Clonetics™ primary cells.

Choose this endothelial cell growth media for an easy cultivation of primary endothelial cells. The SingleQuots™ EGM™-2MV kit is supplied with growth factors, cytokines and supplements, and does not include the basal medium.

  • For coronary and iliac artery, human microvascular endothelial cells
  • Contains 5% FBS and VEGF for rapid proliferation
  • Does not contain Bovine Brain Extract (BBE)
  • Final serum concentration is increased to 5%

Order Info

dimProductType

TRUSTED_SUSTAINABILITY

Specifications

Product Type Growth Medium
Without Additives No Bovine Brain Extract (BBE), No Basal Medium
For Use With (Application) Cultivation of coronary and iliac artery, human microvascular endothelial cells
Quantity 1 Kit
Content And Storage Contains:
  • hEGF
  • Hydrocortisone
  • GA-1000 (Gentamicin, Amphotericin-B)
  • FBS (Fetal Bovine Serum) 25ml
  • VEGF
  • hFGF-B
  • R3-IGF-1
  • Ascorbic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.