missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Endothelin 3 Monoclonal Antibody (3B6)

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561573

Product Code. 30535535

  • 379.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

The protein encoded by this gene is a member of the endothelin family. Endothelins are endothelium-derived vasoactive peptides involved in a variety of biological functions. The active form of this protein is a 21 amino acid peptide processed from the precursor protein. The active peptide is a ligand for endothelin receptor type B (EDNRB). The interaction of this endothelin with EDNRB is essential for development of neural crest-derived cell lineages, such as melanocytes and enteric neurons. Mutations in this gene and EDNRB have been associated with Hirschsprung disease (HSCR) and Waardenburg syndrome (WS), which are congenital disorders involving neural crest-derived cells. Four alternatively spliced transcript variants encoding three distinct isoforms have been observed.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Endothelin 3
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
P14138
EDN3
Human Endothelin-3 synthetic peptide.
100 μg
Primary
Human
Antibody
IgG1
Immunocytochemistry
3B6
Unconjugated
EDN3
114CH19; EDN3; endothelin 3; endothelin-3; ET 3; Et3; ET-3; HSCR4; lethal spotting; ls; MGC15067; MGC61498; PPET3; Preproendothelin 3; preproendothelin-3; preproET-3; RGD1564825; RP4-614C15.1; tmgc48; WS4B
Mouse
Protein A/G
RUO
1908
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.