Learn More
Invitrogen™ Endothelin B Receptor Recombinant Rabbit Monoclonal Antibody (HL2832)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556695
Product Code. 30274986
  • 500.00 € / 100µL
Description
Endothelin B Receptor Recombinant Monoclonal Antibody for IHC (P)
Non-specific receptor for endothelin 1, 2, and 3. Mediates its action by association with G proteins that activate a phosphatidylinositol-calcium second messenger system.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Endothelin B Receptor
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
P21451, P24530, P48302
EDNRB
Synthetic peptide encompassing a sequence within the Intracellular domain of Rat Endothelin B Receptor. The exact sequence is proprietary.
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Immunohistochemistry (Paraffin)
HL2832
Unconjugated
EDNRB
ABCDS; AU022549; Ednra; Ednrb; endothelin B receptor; endothelin receptor non-selective type; endothelin receptor subtype B1; endothelin receptor type B; endothelin-B receptor; ETb; ET-B; Etb receptor; Et-b receptor; ETB1; ETBR; ET-BR; Et-b-r; Etb-svr; ETRB; ET-RB; ETR-b; HSCR; HSCR2; piebald; RP11-318G21.1; Sox10m1; WS4A
Rabbit
Protein A
RUO
13618, 1910, 50672
-20°C, Avoid Freeze/Thaw Cycles