missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enolase 1 Antibody (253), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP2-25147
This item is not returnable.
View return policy
Description
Enolase 1 Monoclonal specifically detects Enolase 1 in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoassay.
Specifications
| Enolase 1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:5000-1:10000, Simple Western 1:500, Immunohistochemistry 1:2000-1:5000, Immunocytochemistry/Immunofluorescence 1:2000-1:5000, Immunoassay | |
| P06733 | |
| ENO1 | |
| The N-terminal 12 amino acids of bovine Enolase 1, which was synthesized on a 8-amine lysine core. [UniProt# Q9XSJ4] | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Bovine, Mouse, Rat, Xenopus. Other species have not been tested. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoassay | |
| 253 | |
| Unconjugated | |
| 50% PBS, 50% glycerol with 5mM Sodium Azide | |
| 2-phospho-D-glycerate hydro-lyase, Alpha enolase, C-myc promoter-binding protein, c-myc promoter-binding protein-1, EC 4.2.1, EC 4.2.1.11, ENO1L1, Enolase 1, enolase 1, (alpha), MBP-1, MBPB1, MPB-1, MPB1c-myc promoter-binding protein 1, MYC promoter-binding protein 1, NNE, Non-neural enolase, Phosphopyruvate hydratase, Plasminogen-binding protein, PPHalpha enolase like 1, tau-crystallin | |
| Mouse | |
| 47 kDa | |
| 0.1 mL | |
| Cancer, Core ESC Like Genes, Stem Cell Markers | |
| 2023 | |
| Human, Mouse, Rat, Pig, Bovine, Equine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction