Learn More
eNOS Antibody (6H2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
Brand:  Novus Biologicals NBP1-51582SS
 View more versions of this product
Product Code. 18701544
  • 213.00 € / 25µL
Description
eNOS Monoclonal specifically detects eNOS in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
eNOS
Monoclonal
1.0 mg/mL
Western Blot 2 ug/ml, ELISA 1:10000, Immunohistochemistry 1:100, Immunohistochemistry-Paraffin 1:100
Constitutive NOS, EC 1.14.13.39, ECNOS, endothelial, Endothelial NOS, eNOScNOS, nitric oxide synthase 3 (endothelial cell), NOS type III, NOSIII
Mouse
133 kDa
0.025 mL
Primary
Human
Precipitation
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
6H2
Unconjugated
P29474
NOS3
Purified recombinant fragment of human eNOS expressed in E. coli. [UniProt# P29474]
Ammonium sulfate precipitation
RUO
4846
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1