missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ ENPP-3/CD203c Antibody (Ags-16C3F) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-28126-100ug
This item is not returnable.
View return policy
Description
ENPP-3/CD203c Monoclonal antibody specifically detects ENPP-3/CD203c in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| ENPP-3/CD203c | |
| Monoclonal | |
| Unconjugated | |
| O14638 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Ags-16C3F | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| CD203c, CD203c antigen, dJ1005H11.3 (phosphodiesterase I/nucleotide pyrophosphatase 3), dJ914N13.3 (phosphodiesterase I/nucleotide pyrophosphatase 3), ectonucleotide pyrophosphatase/phosphodiesterase 3, ectonucleotide pyrophosphatase/phosphodiesterase family member 3, E-NPP 3, gp130RB13-6, NPP3, PD-IBETA, PDNP3B10, Phosphodiesterase I beta, Phosphodiesterase I/nucleotide pyrophosphatase 3, phosphodiesterase-I beta | |
| ENPP3/ CD203c | |
| 100 μg | |
| Protein Phosphatase | |
| 5169 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction