Learn More
Neogen Enrichment Pouch
Description
- Gamma irradiated, ready-to-use pouch with 225 mL Buffered Peptone Water Broth requires no mixing or sterilizing.
- Flat bottom, stand-up pouch ensures stability when adding samples.
- Wire tie holds top open for easy sample addition and closes pouch for transportation.
- Multi-laminate polyester pouch is puncture resistant.
- 2 year shelf life from date of manufacture provides long-term readiness.
Specifications
Specifications
| Test Type | Environmental Test |
| Includes | Buffered Peptone Water Broth |
| Quantity | 32/Cs. |