missing translation for 'onlineSavingsMsg'
Learn More

Entecavir, TRC

Product Code. 30003877
Change view
Click to view available options
Quantity:
10 mg
50 mg
250 mg
missing translation for 'unitSize':
10mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
30003877 250 mg 250mg
30008729 50 mg 50mg
30010859 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30003877 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCE558900250MG
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142217-69-4
Molecular Formula C12 H15 N5 O3
Synonym 6H-Purin-6-one, 2-amino-1,9-dihydro-9-[(1S,3R,4S)-4-hydroxy-3-(hydroxymethyl)-2-methylenecyclopentyl]-∣6H-Purin-6-one, 2-amino-1,9-dihydro-9-[4-hydroxy-3-(hydroxymethyl)-2-methylenecyclopentyl]-, [1S-(1α,3α,4β)]-∣BMS 200475∣Baraclude∣Entecavir∣SQ 34676

Specifications

Format Neat
Molecular Weight (g/mol) 277.28
Product Type API
Shipping Condition Room Temperature
Purity >95% (HPLC)
Quantity 250 mg
Chemical Name or Material Entecavir
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.