missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Oxoid™ Enterobacteria Enrichment Broth-Mossel
Description
Bottled media for Enterobacteria enrichment media complying with EP/USP/JP/BP specification.
Specifications
Specifications
| ATCC® | ARTG |
| Certifications/Compliance | EP/USP/JP/BP |
| Color | Green |
| Content And Storage | Ambient |
| Format | 125 mL sirop bottle |
| Manufacturing Origin | United Kingdom |
| Product Type | Broth |
| Fill Weight | 100-101.5 g |
| Packaging Type | 10 sirop bottles with screw cap closures in a box. |
| Quantity | 10 x 100 mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?