Learn More
Enterokinase Cleavage Site Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA130443
Description
PA1-30443 detects ECS from rabbit samples. PA1-30443 has been successfully used in immunocytochemistry, immunohistochemistry, Western blot applications. The PA1-30443 immunogen is: Rabbits were immunized with ECS enterokinase cleavage site (xxxDDDDK). Antibody was isolated by affinity chromatography using the peptide immobilized on solid support.
Specifications
| Enterokinase Cleavage Site | |
| Polyclonal | |
| Unconjugated | |
| Rabbit | |
| Purified | |
| RUO | |
| Rabbit | |
| Antibody | |
| IgG |
| ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| ECS; ECS Cleavage Site | |
| Rabbits were immunized with ECS enterokinase cleavage site (xxxDDDDK). Antibody was isolated by affinity chromatography using the peptide immobilized on solid support. | |
| 100 μg | |
| Primary | |
| 4° C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.