missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Enteropeptidase/Enterokinase Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00 € - 326.00 €
Specifications
| Antigen | Enteropeptidase/Enterokinase |
|---|---|
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18650392
|
Novus Biologicals
NBP2-95274-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18600255
|
Novus Biologicals
NBP2-95274-50ul |
50 μg |
216.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Enteropeptidase/Enterokinase Polyclonal antibody specifically detects Enteropeptidase/Enterokinase in Bovine samples. It is validated for ELISASpecifications
| Enteropeptidase/Enterokinase | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Epitope Tags, Immunology, Innate Immunity | |
| 0.2 um filtered solution in PBS | |
| 5651 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| ELISA 1:5000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Bovine | |
| EC 3.4.21, EC 3.4.21.9, Enterokinase, ENTKenterokinase, MGC133046, protease, serine, 7 (enterokinase), PRSS7enteropeptidase, Serine protease 7, Transmembrane protease serine 15, transmembrane protease, serine 15 | |
| Produced in rabbits immunized with purified, recombinant Bovine Enteropeptidase/Enterokinase (Accession#: NP_776864.1; Ile801-His1035). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title