Learn More
Neogen Petrifilm™ Environmental Listeria Plate
Description
- Proven cost effective tool for the detection of environmental Listeria.
- Unique results (qualitative, semi-quantitative and quantitative) allow identification of hot spots in plant and track progress against Listeria over time.
- Rapid results in 26-30 hours from sample collection.
- This plate has been awarded Performance Tested Method (PTM) #030601 for stainless steel, ceramic tile and sealed concrete
- Food processors and contract labs worldwide trust Neogen Petrifilm Plates to provide accurate, efficient, consistent results.
Specifications
Specifications
| Content And Storage | Temperatures less than or equal to 8°C |
| Test Type | Environmental Test |
| Quantity | 200 |