Learn More
3M™ Petrifilm™ Environmental Listeria Plate
Detect the presence of Listeria in environmental samples. A sample ready culture medium containing selective agents, nutrients, a cold water soluble gelling agent and a chromogenic indicator.
Brand:  3M™ Petrifilm™ 7100090324
Additional Details : CAS Number : 9000-30-0 Weight : 0.81300kg
 View more versions of this product
Product Code. 17283774
  • 256.52 € / Pack of 50
Estimated Shipment: 15-08-2024
to see stock.
Add to basket
Description
Description
  • Proven cost effective tool for the detection of environmental Listeria.
  • Unique results (qualitative, semi-quantitative and quantitative) allow identification of hot spots in plant and track progress against Listeria over time.
  • Rapid results in 26-30 hours from sample collection.
  • This plate has been awarded Performance Tested Method (PTM) #030601 for stainless steel, ceramic tile and sealed concrete
  • Food processors and contract labs worldwide trust 3M Petrifilm Plates to provide accurate, efficient, consistent results.
Specifications
Specifications
Temperatures less than or equal to 8°C
Food and Beverage, Environmental Test
50