Learn More
3M™ Petrifilm™ Environmental Listeria Plate
Detect the presence of Listeria in environmental samples. A sample ready culture medium containing selective agents, nutrients, a cold water soluble gelling agent and a chromogenic indicator.
256.52 € - 1210.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17273774
|
3M™ Petrifilm™
7100090323 |
200 |
1210.00 €
Pack of 200 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
17283774
|
3M™ Petrifilm™
7100090324 |
50 |
256.52 €
Pack of 50 |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
Description
- Proven cost effective tool for the detection of environmental Listeria.
- Unique results (qualitative, semi-quantitative and quantitative) allow identification of hot spots in plant and track progress against Listeria over time.
- Rapid results in 26-30 hours from sample collection.
- This plate has been awarded Performance Tested Method (PTM) #030601 for stainless steel, ceramic tile and sealed concrete
- Food processors and contract labs worldwide trust 3M Petrifilm Plates to provide accurate, efficient, consistent results.
Specifications
Temperatures less than or equal to 8°C |