missing translation for 'onlineSavingsMsg'
Learn More

GVS Environmental Surface Collection Kit

Product Code. 32178651
Change view
Click to view available options
Media Type:
Buffered Peptone
Butterfields Solution
D-E Neutralizing Broth
Letheen Broth
PBS
Peptone
Saline
Includes:
Attached Foam Swab
Separate Foam Swab
Unit Size:
Pack of 400
14 product options available for selection
Product selection table with 14 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Media Type Includes Unit Size
32178651 D-E Neutralizing Broth Attached Foam Swab Pack of 400
32177662 Butterfields Solution Separate Foam Swab Pack of 400
32177963 Peptone Separate Foam Swab Pack of 400
32178628 PBS Attached Foam Swab Pack of 400
32178026 Butterfields Solution Attached Foam Swab Pack of 400
32178021 Saline Attached Foam Swab Pack of 400
32178109 Buffered Peptone Attached Foam Swab Pack of 400
32178391 Letheen Broth Attached Foam Swab Pack of 400
32178520 Peptone Attached Foam Swab Pack of 400
32178627 D-E Neutralizing Broth Separate Foam Swab Pack of 400
32177776 PBS Separate Foam Swab Pack of 400
32177617 Saline Separate Foam Swab Pack of 400
32177562 Buffered Peptone Separate Foam Swab Pack of 400
32177698 Letheen Broth Separate Foam Swab Pack of 400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 14 options available.
14 options
This item is not returnable. View return policy
Product Code. 32178651 Supplier GVS Supplier No. ESCKC2118X1656A
  • ONLINE EXCLUSIVE PRICE
    508.00 €
    460.88 € / Pack of 400
    Save 47.12 €
    9% Off
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

GVS Environmental surface collection kits available in 10 mL fill volumes with a high-quality foam swab.

  • Available in 10 mL fill volumes
  • With high-quality foam swab
  • Swab can be securely attached to the cap and makes a convenient 'handle' for great control when sampling

Specifications

Appearance Purple to Yellow/Cloudiness
Broth Type D/E Neutralizing Broth
Color Purple to Yellow
Detects Bactericidal or Bacteriostatic Activity of Antiseptic and Disinfectant Chemicals
Format Kit, Tube, Foam Swab
Media Type D-E Neutralizing Broth
Product Type Prepared Microbiology Tubes
Sterility Non-sterile
Test Type Environmental Test
Fill Volume 10 mL
Formulation Neutralizing Agents
For Use With (Application) Environmental Sampling for Food, Pharmaceutical, Biotechnology, and Cosmetic Industries
Includes Attached Foam Swab
Material Foam Swab
No. per Pack 50
Packaging Type Rack/Inner Box, Case
Quantity 10 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.