missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EOGT/AER61 Antibody (S02-0E5), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19559-100ul
This item is not returnable.
View return policy
Description
EOGT/AER61 Monoclonal antibody specifically detects EOGT/AER61 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| EOGT/AER61 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| S02-0E5 | |
| Unconjugated | |
| AER61, AER61 glycosyltransferase, chromosome 3 open reading frame 64, EC 2.4, FLJ13078, FLJ33770, FLJ41219, MGC34132 | |
| Recombinant protein of human EOGT/AER61 (Uniprot # Q5NDL2) | |
| 100 μg | |
| Proteases & Other Enzymes | |
| 285203 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction