missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EOMES Antibody (CL15426) [Unconjugated], Novus Biologicals™
Mouse Monoclonal Antibody
422.00 € - 605.00 €
Specifications
| Antigen | EOMES |
|---|---|
| Clone | CL15426 |
| Concentration | 1 mg/mL |
| Dilution | Immunohistochemistry 1:200-1:500, Immunohistochemistry-Paraffin 1:200-1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30279821
|
Novus Biologicals
NBP3-43668-100ul |
100 μL |
605.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30279722
|
Novus Biologicals
NBP3-43668-25ul |
25 μL |
422.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EOMES Monoclonal antibody specifically detects EOMES in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)Specifications
| EOMES | |
| 1 mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Innate Immunity | |
| PBS, pH 7.2, 40% glycerol | |
| 8320 | |
| IgG1 | |
| Protein A purified |
| CL15426 | |
| Immunohistochemistry 1:200-1:500, Immunohistochemistry-Paraffin 1:200-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| eomesodermin, eomesodermin (Xenopus laevis) homolog, t box, brain, 2, T-box brain protein 2, T-box brain2, TBR-2, TBR2eomesodermin homolog, T-brain-2 | |
| This antibody was generated using a recombinant protein sequence of O95936, with the exact immunogen sequence remaining proprietary. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title