missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Eotaxin-3-like Polyclonal Antibody
GREENER_CHOICE

Product Code. 30535011
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30535011 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30535011 Supplier Invitrogen™ Supplier No. PA5148769

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

Eotaxin-3-like, also known as CCL26 (C-C motif chemokine ligand 26), is a member of the eotaxin family of chemokines, which play crucial roles in the immune system by mediating the recruitment and activation of eosinophils. Eotaxin-3-like is primarily expressed in epithelial cells and is upregulated in response to inflammatory signals such as interleukin-4 (IL-4) and interleukin-13 (IL-13). This chemokine binds to the CCR3 receptor on eosinophils, promoting their migration to sites of inflammation or allergic reactions. Eotaxin-3-like is particularly important in conditions such as asthma, allergic rhinitis, and atopic dermatitis, where elevated levels contribute to the accumulation of eosinophils and the resulting inflammatory response. It is also implicated in other diseases characterized by eosinophilic infiltration, such as eosinophilic esophagitis. Understanding the regulation and function of eotaxin-3-like is essential for developing targeted therapies aimed at modulating eosinophilic responses and treating allergic and inflammatory diseases. Research into eotaxin-3-like provides valuable insights into the broader mechanisms of chemokine-mediated immune cell trafficking and its potential as a therapeutic target for controlling eosinophil-related pathologies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Eotaxin-3-like
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with trehalose and no preservative
Host Species Goat
Immunogen E. coli-derived recombinant rat CCL26-like/Eotaxin-3-like, Thr24-Trp94.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.