missing translation for 'onlineSavingsMsg'
Learn More

EPA 8270 B/N Surrogate Mixture 1000 ug/mL in Dichloromethane, Dr. Ehrenstorfer

Product Code. 30319422
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30319422 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30319422 Supplier Dr. Ehrenstorfer Supplier No. DREYA09000007DI
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Fluorobiphenyl∣Nitrobenzene D5∣p-Terphenyl D14
CAS 321-60-8, 4165-60-0, 1718-51-0
Analyte or Component Names 2-Fluorobiphenyl∣Nitrobenzene D5∣p-Terphenyl D14
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dichloromethane
Industry Type Food Testing
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 2-Fluorobiphenyl 1000 μg/mL∣Nitrobenzene D5 1000 μg/mL∣p-Terphenyl D14 1000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.