Learn More
Beskrivning
Innehåller
Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
| Analyte | CAS No. | Concentration |
|---|---|---|
| 1,2-Dibromo-3-chloropropane | 96-12-8 | 2000μg/mL |
| 1,2-Dibromoethane | 106-93-4 | 2000μg/mL |
| 1,2-Dichlorobenzene | 95-50-1 | 2000μg/mL |
| 1,3-Dichlorobenzene | 541-73-1 | 2000μg/mL |
| 1,4-Dichlorobenzene | 106-46-7 | 2000μg/mL |
| Bromochloromethane | 74-97-5 | 2000μg/mL |
Certifikat
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Uppfyllande
GHS/OSHA Complaint
Specifikationer
Specifikationer
| Chemical Name or Material | EPA CLP Volatiles Standards |
| Packaging | 2mL amber prescored glass ampule |
| CAS | 106-46-7, 7732-18-5, 106-93-4, 95-50-1, 541-73-1 |
| Concentration or Composition Notes | Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
| Methods | For US EPA CLP Methods |
| Analyte or Component Names | 1,2-Dibromo-3-chloropropane,1 |
| For Use With (Equipment) | GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
| Element | 1,2-Dibromo-3-chloropropane |
| Solvent or Matrix | Methanol-P&T |
| Quantity | 1 mL |
| Visa mer |
Säkerhet och hantering
- EPA CLP Volatiles Standards
- Danger
- Acute toxicity Category 3
- Carcinogenicity Category 1B
- Flammable liquid Category 2
- Germ cell mutagenicity Category 1B
- Reproductive toxicity Category 1
- Specific target organ toxicity after single exposure Category 1
- H225-Highly flammable liquid and vapour.
- H331-Toxic if inhaled.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H360-May damage fertility or the unborn child.
- H370-Causes damage to organs.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contain: methanol, 1,2-dibromo-3-chloropropane, 1,4-dichlorobenzene,1,2-dibromoethane
Product intended for laboratory use only
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.