missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 1631, 1N Bromide/1N Bromate in Deionized Water, 10 x 10 mL, smart delivery PFA tubes, Dr. Ehrenstorfer

Product Code. 30308797
Change view
Click to view available options
Quantity:
10 x 10 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30308797 10 x 10 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30308797 Supplier Dr. Ehrenstorfer Supplier No. DREO000003
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium Bromate∣Potassium bromide
CAS 1-2-7758, 2-3-7758
Analyte or Component Names Potassium Bromate∣Potassium bromide
Shipping Condition Room Temperature
Certifications/Compliance ISO 17025
Solvent or Matrix Water
Industry Type Environmental Testing
Quantity 10 x 10 mL
Concentration or Composition (by Analyte or Components) Potassium Bromate 1 N∣Potassium bromide 1 N
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.