missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA Method 200.8: 100 mg/L Ba, Cu, Fe, Zn; 10 mg/L Ag, 100mLs, VHG™
Description
EPA Method 200.8: 100 mg/L Ba, Cu, Fe, Zn; 10 mg/L Ag, 100mLs
Specifications
Specifications
| Chemical Name or Material | Silver/Barium/Copper/Iron/Zinc |
| CAS | 7440-22-4, 7440-39-3, 7440-50-8, 7439-89-6, 7440-66-6 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Silver/Barium/Copper/Iron/Zinc |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dilute HNO3 |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Silver 10 mg/L/Barium 100 mg/L/Copper 100 mg/L/Iron 100 mg/L/Zinc 100 mg/L |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?