missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 200.8: 100 mg/L Sc, Y, In, Tb, Bi, 100mls , VHG™

Product Code. 17958319
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17958319 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17958319 Supplier VHG Labs Supplier No. VHG1600685
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

EPA Method 200.8: 100 mg/L Sc, Y, In, Tb, Bi, 100mls
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bismuth/Indium/Scandium/Terbium/Yttrium
CAS 7440-69-9, 7440-74-6, 7440-20-2, 7440-27-9, 7440-65-5
Methods ICP/ICP-MS
Analyte or Component Names Bismuth/Indium/Scandium/Terbium/Yttrium
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Dilute HNO3
Quantity 100 mL
Concentration or Composition (by Analyte or Components) Bismuth 100 mg/L/Indium 100 mg/L/Scandium 100 mg/L/Terbium 100 mg/L/Yttrium 100 mg/L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.