missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA Method 200.8: SS-10 mg/L Al, As, Be, Cd, Co, Cr, Mn, Mo, Ni, Pb, Sb, Se, Tl, Th, U, V. Made fro, VHG™
Description
EPA Method 200.8: SS-10 mg/L Al, As, Be, Cd, Co, Cr, Mn, Mo, Ni, Pb, Sb, Se, Tl, Th, U, V. Made fro
Specifications
Specifications
| Chemical Name or Material | Aluminium/Antimony/Arsenic/Beryllium/Cadmium/Chromium/Cobalt/Lead/Manganese/Molybdenum/Nickel/Selenium/Thallium/Thorium/Uranium/Vanadium |
| CAS | 7429-90-5, 7440-36-0, 7440-38-2, 7440-41-7, 7440-43-9, 7440-47-3 |
| Methods | ICP/ICP-MS |
| Analyte or Component Names | Aluminium/Antimony/Arsenic/Beryllium/Cadmium/Chromium/Cobalt/Lead/Manganese/Molybdenum/Nickel/Selenium/Thallium/Thorium/Uranium/Vanadium |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Dilute HNO3 |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Aluminium 10 mg/L/Antimony 10 mg/L/Arsenic 10 mg/L/Beryllium 10 mg/L/Cadmium 10 mg/L/Chromium 10 mg/L/Cobalt 10 mg/L/Lead 10 mg/L/Manganese 10 mg/L/Molybdenum 10 mg/L/Nickel 10 mg/L/Selenium 10 mg/L/Thallium 10 mg/L/Thorium 10 mg/L/Uranium 10 mg/L/Vanadium 10 mg/L |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?