missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 5035, Low Level Clear Vial, 20% Bisulfate Solution, Stir Bar, Tared, 5 mL, 72/cs, Dr. Ehrenstorfer

Product Code. 30328747
Change view
Click to view available options
Analyte or Component Names:
Sodium Bisulfite (mixture of NaHSO3 and Na2S2O5)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Analyte or Component Names unitSize
30328747 Sodium Bisulfite (mixture of NaHSO3 and Na2S2O5) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30328747 Supplier Dr. Ehrenstorfer Supplier No. DREO000012
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sodium Bisulfite (mixture of NaHSO3 and Na2S2O5)
CAS 7631-90-5
Analyte or Component Names Sodium Bisulfite (mixture of NaHSO3 and Na2S2O5)
Shipping Condition Room Temperature
Molecular Formula NaHSO3
Concentration 20 %
Certifications/Compliance ISO 17025
Synonym Sulfurous Acid Monosodium Salt, E 222, FR-62, Hydrogen Sodium Sulfite, Hydrogen Sulfite Sodium, Monosodium Sulfite, Sodium Acid Sulfite, Sodium Bisulfite, Sodium Bisulfite (NaHSO3), Sodium Bisulphite, Sodium Hydrogen Sulfite, Sodium Sulfite (NaHSO3);
SMILES [Na+].OS(=O)[O-]
Industry Type Environmental Testing
Molecular Weight (g/mol) 104.06
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.