missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 505 Stock Standard Mixture 375 100 μg/mL in Toluene, Dr. Ehrenstorfer

Product Code. 30317208
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30317208 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30317208 Supplier Dr. Ehrenstorfer Supplier No. DREA50000375TO
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acenaphthene∣Acenaphthylene∣Anthracene∣Benz[a]anthracene∣Benzo(k)fluoranthene∣Benzo[a]pyrene∣Benzo[b]fluoranthene∣Benzo[ghi]perylene∣Chrysene∣Dibenzo[a,c]anthracene∣Fluoranthene∣Fluorene∣Indeno[1,2,3-cd]pyrene∣Naphthalene∣Phenanthrene∣Pyrene
CAS 83-32-9, 208-96-8, 120-12-7, 56-55-3, 207-08-9, 50-32-8
Analyte or Component Names Acenaphthene∣Acenaphthylene∣Anthracene∣Benz[a]anthracene∣Benzo(k)fluoranthene∣Benzo[a]pyrene∣Benzo[b]fluoranthene∣Benzo[ghi]perylene∣Chrysene∣Dibenzo[a,c]anthracene∣Fluoranthene∣Fluorene∣Indeno[1,2,3-cd]pyrene∣Naphthalene∣Phenanthrene∣Pyrene
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Toluene
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Acenaphthene 100 μg/mL∣Acenaphthylene 100 μg/mL∣Anthracene 100 μg/mL∣Benz[a]anthracene 100 μg/mL∣Benzo(k)fluoranthene 100 μg/mL∣Benzo[a]pyrene 100 μg/mL∣Benzo[b]fluoranthene 100 μg/mL∣Benzo[ghi]perylene 100 μg/mL∣Chrysene 100 μg/mL∣Dibenzo[a,c]anthracene 100 μg/mL∣Fluoranthene 100 μg/mL∣Fluorene 100 μg/mL∣Indeno[1,2,3-cd]pyrene 100 μg/mL∣Naphthalene 100 μg/mL∣Phenanthrene 100 μg/mL∣Pyrene 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.