missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 515.2 Herbicide Mixture 402 100-1000 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30324754
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30324754 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324754 Supplier Dr. Ehrenstorfer Supplier No. DREA50000402ME
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4-Dichlorophenoxyacetic acid∣4-(2,4-Dichlorophenoxy)butyric acid∣Acifluorfen∣Bentazone∣Dicamba∣Fenoprop∣Picloram
CAS 94-75-7, 94-82-6, 50594-66-6, 25057-89-0, 1918-00-9, 93-72-1
Analyte or Component Names 2,4-Dichlorophenoxyacetic acid∣4-(2,4-Dichlorophenoxy)butyric acid∣Acifluorfen∣Bentazone∣Dicamba∣Fenoprop∣Picloram
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 2,4-Dichlorophenoxyacetic acid 100 μg/mL∣Fenoprop 100 μg/mL∣Acifluorfen 200 μg/mL∣Dicamba 300 μg/mL∣Picloram 300 μg/mL∣4-(2,4-Dichlorophenoxy)butyric acid 1000 μg/mL∣Bentazone 1000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.