missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA Method 515.3 Herbicide Mixture 405 10-100 μg/mL in Acetone, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | 2,4,5-Trichlorophenoxyacetic acid∣2,4-Dichlorophenoxyacetic acid∣3,5-Dichlorobenzoic acid∣4-(2,4-Dichlorophenoxy)butyric acid∣4-Nitrophenol∣Acifluorfen∣Bentazone∣Chloramben∣Chlorthal-diacid∣Dacthal monoacid∣Dalapon∣Dicamba∣Dichlorprop∣Dinoseb∣Fenoprop∣Pentachlorophenol∣Picloram |
| CAS | 93-76-5, 94-75-7, 51-36-5, 94-82-6, 100-02-7, 50594-66-6 |
| Analyte or Component Names | 2,4,5-Trichlorophenoxyacetic acid∣2,4-Dichlorophenoxyacetic acid∣3,5-Dichlorobenzoic acid∣4-(2,4-Dichlorophenoxy)butyric acid∣4-Nitrophenol∣Acifluorfen∣Bentazone∣Chloramben∣Chlorthal-diacid∣Dacthal monoacid∣Dalapon∣Dicamba∣Dichlorprop∣Dinoseb∣Fenoprop∣Pentachlorophenol∣Picloram |
| Shipping Condition | +20°C |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetone |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Pentachlorophenol 10 μg/mL∣2,4,5-Trichlorophenoxyacetic acid 25 μg/mL∣Fenoprop 25 μg/mL∣3,5-Dichlorobenzoic acid 50 μg/mL∣Acifluorfen 50 μg/mL∣Chloramben 50 μg/mL∣Chlorthal-diacid 50 μg/mL∣Dacthal monoacid 50 μg/mL∣Dicamba 50 μg/mL∣2,4-Dichlorophenoxyacetic acid 100 μg/mL∣4-(2,4-Dichlorophenoxy)butyric acid 100 μg/mL∣4-Nitrophenol 100 μg/mL∣Bentazone 100 μg/mL∣Dalapon 100 μg/mL∣Dichlorprop 100 μg/mL∣Dinoseb 100 μg/mL∣Picloram 100 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?