missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 515.4 Herbicide Mixture 409 10-100 μg/mL in Methyl tert Butyl Ether, Dr. Ehrenstorfer

Product Code. 30309227
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30309227 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30309227 Supplier Dr. Ehrenstorfer Supplier No. DREA50000409MB
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4,5-T-methyl ester∣2,4-DB-methyl ester∣2,4-D-methyl ester∣3,5-Dichlorobenzoic acid methyl ester∣Acifluorfen methyl ester∣Bentazone-methyl∣Chloramben methyl ester∣Chlorthal-dimethyl∣Dalapon-methyl ester∣Dicamba-methyl ester∣Dichlorprop-methyl ester∣Dinoseb methyl ether∣Fenoprop-methyl ester∣Pentachloroanisole∣Picloram-methyl ester∣Quinclorac-methyl ester
CAS 1928-37-6, 18625-12-2, 1928-38-7, 2905-67-1, 50594-67-7, 61592-45-8
Analyte or Component Names 2,4,5-T-methyl ester∣2,4-DB-methyl ester∣2,4-D-methyl ester∣3,5-Dichlorobenzoic acid methyl ester∣Acifluorfen methyl ester∣Bentazone-methyl∣Chloramben methyl ester∣Chlorthal-dimethyl∣Dalapon-methyl ester∣Dicamba-methyl ester∣Dichlorprop-methyl ester∣Dinoseb methyl ether∣Fenoprop-methyl ester∣Pentachloroanisole∣Picloram-methyl ester∣Quinclorac-methyl ester
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix MTBE
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Pentachloroanisole 10 μg/mL∣2,4,5-T-methyl ester 25 μg/mL∣Fenoprop-methyl ester 25 μg/mL∣3,5-Dichlorobenzoic acid methyl ester 50 μg/mL∣Acifluorfen methyl ester 50 μg/mL∣Chloramben methyl ester 50 μg/mL∣Dicamba-methyl ester 50 μg/mL∣Picloram-methyl ester 50 μg/mL∣Quinclorac-methyl ester 50 μg/mL∣2,4-DB-methyl ester 100 μg/mL∣2,4-D-methyl ester 100 μg/mL∣Bentazone-methyl 100 μg/mL∣Chlorthal-dimethyl 100 μg/mL∣Dalapon-methyl ester 100 μg/mL∣Dichlorprop-methyl ester 100 μg/mL∣Dinoseb methyl ether 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.