missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 524.3 0055 Internal Standard Mixture 2000 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30315309
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30315309 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30315309 Supplier Dr. Ehrenstorfer Supplier No. DREA50000055ME
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,4-Dichlorobenzene-D4∣1,4-Difluorobenzene∣Chlorobenzene D5
CAS 3855-82-1, 540-36-3, 3114-55-4
Analyte or Component Names 1,4-Dichlorobenzene-D4∣1,4-Difluorobenzene∣Chlorobenzene D5
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Methanol
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1,4-Dichlorobenzene-D4 2000 μg/mL∣1,4-Difluorobenzene 2000 μg/mL∣Chlorobenzene D5 2000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.