missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPA Method 525.1 PAH Mixture 384 100 μg/mL in Acetone, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Acenaphthylene∣Anthracene∣Benz[a]anthracene∣Benzo(j)fluoranthene∣Benzo(k)fluoranthene∣Benzo[a]pyrene∣Benzo[ghi]perylene∣Chrysene∣Dibenzo(a,h)anthracene∣Fluorene∣Indeno[1,2,3-cd]pyrene∣Phenanthrene∣Pyrene |
| CAS | 208-96-8, 120-12-7, 56-55-3, 205-82-3, 207-08-9, 50-32-8 |
| Analyte or Component Names | Acenaphthylene∣Anthracene∣Benz[a]anthracene∣Benzo(j)fluoranthene∣Benzo(k)fluoranthene∣Benzo[a]pyrene∣Benzo[ghi]perylene∣Chrysene∣Dibenzo(a,h)anthracene∣Fluorene∣Indeno[1,2,3-cd]pyrene∣Phenanthrene∣Pyrene |
| Shipping Condition | Room Temperature |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Acetone |
| Quantity | 1 mL |
| Concentration or Composition (by Analyte or Components) | Acenaphthylene 100 μg/mL∣Anthracene 100 μg/mL∣Benz[a]anthracene 100 μg/mL∣Benzo(j)fluoranthene 100 μg/mL∣Benzo(k)fluoranthene 100 μg/mL∣Benzo[a]pyrene 100 μg/mL∣Benzo[ghi]perylene 100 μg/mL∣Chrysene 100 μg/mL∣Dibenzo(a,h)anthracene 100 μg/mL∣Fluorene 100 μg/mL∣Indeno[1,2,3-cd]pyrene 100 μg/mL∣Phenanthrene 100 μg/mL∣Pyrene 100 μg/mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction