missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 525.1 PAH Mixture 385 500 μg/mL in Toluene, Dr. Ehrenstorfer

Product Code. 30321025
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30321025 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30321025 Supplier Dr. Ehrenstorfer Supplier No. DREA50000385TO
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Acenaphthylene∣Anthracene∣Benz[a]anthracene∣Benzo(j)fluoranthene∣Benzo(k)fluoranthene∣Benzo[a]pyrene∣Benzo[ghi]perylene∣Chrysene∣Dibenzo(a,h)anthracene∣Fluorene∣Indeno[1,2,3-cd]pyrene∣Phenanthrene∣Pyrene
CAS 208-96-8, 120-12-7, 56-55-3, 205-82-3, 207-08-9, 50-32-8
Analyte or Component Names Acenaphthylene∣Anthracene∣Benz[a]anthracene∣Benzo(j)fluoranthene∣Benzo(k)fluoranthene∣Benzo[a]pyrene∣Benzo[ghi]perylene∣Chrysene∣Dibenzo(a,h)anthracene∣Fluorene∣Indeno[1,2,3-cd]pyrene∣Phenanthrene∣Pyrene
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Toluene
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Acenaphthylene 500 μg/mL∣Anthracene 500 μg/mL∣Benz[a]anthracene 500 μg/mL∣Benzo(j)fluoranthene 500 μg/mL∣Benzo(k)fluoranthene 500 μg/mL∣Benzo[a]pyrene 500 μg/mL∣Benzo[ghi]perylene 500 μg/mL∣Chrysene 500 μg/mL∣Dibenzo(a,h)anthracene 500 μg/mL∣Fluorene 500 μg/mL∣Indeno[1,2,3-cd]pyrene 500 μg/mL∣Phenanthrene 500 μg/mL∣Pyrene 500 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.