missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 525.3 Organochlorine Pesticide Mixture 500 μg/mL in Acetone, Dr. Ehrenstorfer

Product Code. 30324636
Click to view available options
Quantity:
5 x 1 mL
Unit Size:
5mL
This item is not returnable. View return policy

Product Code. 30324636

Brand: Dr. Ehrenstorfer DREGS09000341AC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (+)-trans-Permethrin∣1,3-Phenylenediamine∣4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Acetochlor∣Aldrin∣alpha-Endosulfan∣alpha-HCH∣beta-Endosulfan∣beta-HCH∣Chlorobenzilate∣Chloroneb∣Chlorothalonil∣Chlorthal-dimethyl∣Cis-Chlordane (Alpha Isomer)∣cis-Permethrin (mix of isomers)∣delta-HCH∣Dieldrin∣Endosulfan-sulfate∣Endrin∣Heptachlor∣Heptachlor-exo-epoxide (cis-, isomer B)∣Hexachlorobenzene∣Hexachlorocyclopentadiene∣Methoxychlor∣Pentachlorophenol∣Trans-Chlordane (Gamma Isomer)∣trans-Nonachlor
CAS 51877-74-8, 108-45-2, 72-54-8, 72-55-9, 50-29-3, 34256-82-1
Analyte or Component Names (+)-trans-Permethrin∣1,3-Phenylenediamine∣4,4'-DDD∣4,4'-DDE∣4,4'-DDT∣Acetochlor∣Aldrin∣alpha-Endosulfan∣alpha-HCH∣beta-Endosulfan∣beta-HCH∣Chlorobenzilate∣Chloroneb∣Chlorothalonil∣Chlorthal-dimethyl∣Cis-Chlordane (Alpha Isomer)∣cis-Permethrin (mix of isomers)∣delta-HCH∣Dieldrin∣Endosulfan-sulfate∣Endrin∣Heptachlor∣Heptachlor-exo-epoxide (cis-, isomer B)∣Hexachlorobenzene∣Hexachlorocyclopentadiene∣Methoxychlor∣Pentachlorophenol∣Trans-Chlordane (Gamma Isomer)∣trans-Nonachlor
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Acetone
Quantity 5 x 1 mL
Concentration or Composition (by Analyte or Components) (+)-trans-Permethrin 500 μg/mL∣1,3-Phenylenediamine 500 μg/mL∣4,4'-DDD 500 μg/mL∣4,4'-DDE 500 μg/mL∣4,4'-DDT 500 μg/mL∣Acetochlor 500 μg/mL∣Aldrin 500 μg/mL∣alpha-Endosulfan 500 μg/mL∣alpha-HCH 500 μg/mL∣beta-Endosulfan 500 μg/mL∣beta-HCH 500 μg/mL∣Chlorobenzilate 500 μg/mL∣Chloroneb 500 μg/mL∣Chlorothalonil 500 μg/mL∣Chlorthal-dimethyl 500 μg/mL∣Cis-Chlordane (Alpha Isomer) 500 μg/mL∣cis-Permethrin (mix of isomers) 500 μg/mL∣delta-HCH 500 μg/mL∣Dieldrin 500 μg/mL∣Endosulfan-sulfate 500 μg/mL∣Endrin 500 μg/mL∣Heptachlor 500 μg/mL∣Heptachlor-exo-epoxide (cis-, isomer B) 500 μg/mL∣Hexachlorobenzene 500 μg/mL∣Hexachlorocyclopentadiene 500 μg/mL∣Methoxychlor 500 μg/mL∣Pentachlorophenol 500 μg/mL∣Trans-Chlordane (Gamma Isomer) 500 μg/mL∣trans-Nonachlor 500 μg/mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.