missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 528 Phenol Calibration Mixture 389 2000 μg/mL in Dichloromethane, Dr. Ehrenstorfer

Product Code. 30323100
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30323100 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30323100 Supplier Dr. Ehrenstorfer Supplier No. DREA50000389DI
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2,4,6-Trichlorophenol∣2,4-Dichlorophenol∣2,4-Dimethylphenol∣2,4-Dinitrophenol∣2-Chlorophenol∣2-Methyl-4,6-dinitrophenol∣2-Methylphenol∣2-Nitrophenol∣4-Chloro-3-methylphenol∣4-Nitrophenol∣Pentachlorophenol∣Phenol
CAS 88-06-2, 120-83-2, 105-67-9, 51-28-5, 95-57-8, 534-52-1
Analyte or Component Names 2,4,6-Trichlorophenol∣2,4-Dichlorophenol∣2,4-Dimethylphenol∣2,4-Dinitrophenol∣2-Chlorophenol∣2-Methyl-4,6-dinitrophenol∣2-Methylphenol∣2-Nitrophenol∣4-Chloro-3-methylphenol∣4-Nitrophenol∣Pentachlorophenol∣Phenol
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solvent or Matrix Dichloromethane
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 2,4,6-Trichlorophenol 2000 μg/mL∣2,4-Dichlorophenol 2000 μg/mL∣2,4-Dimethylphenol 2000 μg/mL∣2,4-Dinitrophenol 2000 μg/mL∣2-Chlorophenol 2000 μg/mL∣2-Methyl-4,6-dinitrophenol 2000 μg/mL∣2-Methylphenol 2000 μg/mL∣2-Nitrophenol 2000 μg/mL∣4-Chloro-3-methylphenol 2000 μg/mL∣4-Nitrophenol 2000 μg/mL∣Pentachlorophenol 2000 μg/mL∣Phenol 2000 μg/mL
Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.