missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 533 Isotope Performance Standard, 1-3 μg/mL in Methanol:Water, Dr. Ehrenstorfer

Product Code. 31891582
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31891582 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31891582 Supplier Dr. Ehrenstorfer Supplier No. DREA50000795MW
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Chemical Name or Material Perfluorobutanoic acid 13C3 (2,3,4-13C3),Perfluorooctanesulfonic Acid 13C4 (1,2,3,4-13C4) Sodium,Perfluorooctanoic Acid-13C2
Packaging Bottle
CAS 2483735-33-5, 960315-53-1, 864071-08-9
Concentration or Composition Notes Perfluorobutanoic acid 13C3 (2,3,4-13C3) 1 μg/mL,Perfluorooctanesulfonic Acid 13C4 (1,2,3,4-13C4) Sodium 3 μg/mL,Perfluorooctanoic Acid-13C2 1 μg/mL
Analyte or Component Names Perfluorobutanoic acid 13C3 (2,3,4-13C3),Perfluorooctanesulfonic Acid 13C4 (1,2,3,4-13C4) Sodium,Perfluorooctanoic Acid-13C2
Shipping Condition +20°C
Certifications/Compliance ISO 17034
Solution Type Mixture
Type Multi-component Standard
Solvent or Matrix Methanol/Water
Quantity 1 mL
Grade Ungraded
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.