missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 552 Halogenated Acetic Mixture 100 μg/mL in Methyl tert-butyl ether, Dr. Ehrenstorfer

Product Code. 30314364
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30314364 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30314364 Supplier Dr. Ehrenstorfer Supplier No. DREGA09000361MB
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bromoacetic acid∣Bromochloroacetic acid∣Bromodichloroacetic acid∣Chloroacetic acid∣Dalapon∣Dibromoacetic acid∣Dibromochloroacetic acid∣Dichloroacetic acid∣Tribromoacetic acid∣Trichloroacetic acid (TCA)
CAS 79-08-3, 5589-96-8, 71133-14-7, 79-11-8, 75-99-0, 631-64-1
Analyte or Component Names Bromoacetic acid∣Bromochloroacetic acid∣Bromodichloroacetic acid∣Chloroacetic acid∣Dalapon∣Dibromoacetic acid∣Dibromochloroacetic acid∣Dichloroacetic acid∣Tribromoacetic acid∣Trichloroacetic acid (TCA)
Shipping Condition Ice pack (-20°C)
Certifications/Compliance ISO 17034
Solvent or Matrix MTBE
Quantity 1 mL
Concentration or Composition (by Analyte or Components) Bromoacetic acid 100 μg/mL∣Bromochloroacetic acid 100 μg/mL∣Bromodichloroacetic acid 100 μg/mL∣Chloroacetic acid 100 μg/mL∣Dalapon 100 μg/mL∣Dibromoacetic acid 100 μg/mL∣Dibromochloroacetic acid 100 μg/mL∣Dichloroacetic acid 100 μg/mL∣Tribromoacetic acid 100 μg/mL∣Trichloroacetic acid (TCA) 100 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.