missing translation for 'onlineSavingsMsg'
Learn More

EPA Method 556 Carbonyl Mixture 457 1000 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30311285
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30311285 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30311285 Supplier Dr. Ehrenstorfer Supplier No. DREA50000457AL
 Restricted Item
Estimated Shipment: 25-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Heptanal∣Acetaldehyde∣Benzaldehyde∣Butanal∣Caproic aldehyde∣Crotonaldehyde (Butenal)∣Cyclohexanone∣Decanal∣Formaldehyde∣Glyoxal∣n-Valeraldehyde∣Octanal∣Pelargonaldehyde∣Propionaldehyde∣Pyruvaldehyde
CAS 111-71-7, 75-07-0, 100-52-7, 123-72-8, 66-25-1, 123-73-9
Analyte or Component Names 1-Heptanal∣Acetaldehyde∣Benzaldehyde∣Butanal∣Caproic aldehyde∣Crotonaldehyde (Butenal)∣Cyclohexanone∣Decanal∣Formaldehyde∣Glyoxal∣n-Valeraldehyde∣Octanal∣Pelargonaldehyde∣Propionaldehyde∣Pyruvaldehyde
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Acetonitrile
Quantity 1 mL
Concentration or Composition (by Analyte or Components) 1-Heptanal 1000 μg/mL∣Acetaldehyde 1000 μg/mL∣Benzaldehyde 1000 μg/mL∣Butanal 1000 μg/mL∣Caproic aldehyde 1000 μg/mL∣Crotonaldehyde (Butenal) 1000 μg/mL∣Cyclohexanone 1000 μg/mL∣Decanal 1000 μg/mL∣Formaldehyde 1000 μg/mL∣Glyoxal 1000 μg/mL∣n-Valeraldehyde 1000 μg/mL∣Octanal 1000 μg/mL∣Pelargonaldehyde 1000 μg/mL∣Propionaldehyde 1000 μg/mL∣Pyruvaldehyde 1000 μg/mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.